PloS one
Wnts enhance neurotrophin-induced neuronal differentiation in adult bone-marrow-derived mesenchymal stem cells via canonical and noncanonical signaling pathways.
PMID 25170755
Abstract
Wnts were previously shown to regulate the neurogenesis of neural stem or progenitor cells. Here, we explored the underlying molecular mechanisms through which Wnt signaling regulates neurotrophins (NTs) in the NT-induced neuronal differentiation of human mesenchymal stem cells (hMSCs). NTs can increase the expression of Wnt1 and Wnt7a in hMSCs. However, only Wnt7a enables the expression of synapsin-1, a synaptic marker in mature neurons, to be induced and triggers the formation of cholinergic and dopaminergic neurons. Human recombinant (hr)Wnt7a and general neuron makers were positively correlated in a dose- and time-dependent manner. In addition, the expression of synaptic markers and neurites was induced by Wnt7a and lithium, a glycogen synthase kinase-3β inhibitor, in the NT-induced hMSCs via the canonical/β-catenin pathway, but was inhibited by Wnt inhibitors and frizzled-5 (Frz5) blocking antibodies. In addition, hrWnt7a triggered the formation of cholinergic and dopaminergic neurons via the non-canonical/c-jun N-terminal kinase (JNK) pathway, and the formation of these neurons was inhibited by a JNK inhibitor and Frz9 blocking antibodies. In conclusion, hrWnt7a enhances the synthesis of synapse and facilitates neuronal differentiation in hMSCS through various Frz receptors. These mechanisms may be employed widely in the transdifferentiation of other adult stem cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB2081
Anti-β-Catenin Antibody, clone 5H10, Anti-β-Catenin Antibody, clone 5H10 is a Mouse Monoclonal Antibody for detection of β-Catenin also known as beta catenin & has been tested in ICC, IHC, IHC(P), IP & WB.
MAB305
Anti-Choline Acetyltransferase Antibody, clone 1E6, Detect Choline Acetyltransferase using this Anti-Choline Acetyltransferase Antibody, clone 1E6 validated for use in IH.
AB1585
Anti-Dopamine β Hydroxylase Antibody, Detect Dopamine β Hydroxylase using this Anti-Dopamine β Hydroxylase Antibody validated for use in IH, FUNC, RIA & WB.
ABS16
Anti-GAPDH Antibody, Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody is popular affinity purified polyclonal antibody. This antibody is supported by peer reviewed publications and reliably detects Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) is validated for use in WB.
AB5622
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Detect Microtubule-Associated Protein 2 (MAP2) using this Anti-Microtubule-Associated Protein 2 (MAP2) Antibody validated for use in ELISA, IC, IH, IH(P) & WB with more than 55 product citations.
AB1543
Anti-Synapsin I Antibody, Detect Synapsin I using this Anti-Synapsin I Antibody validated for use in IC, IH(P), IP & WB.
HPA010926
Anti-ALCAM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22783 Anti-CD14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500215
Anti-CD14 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405579
Anti-CD14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001887
Anti-CD14 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002127
Anti-CD14 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306488
ANTI-CD166(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW10811F Anti-CD34 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4300690
Anti-CD34 antibody produced in rabbit, affinity isolated antibody
SAB1402685
Monoclonal Anti-ALCAM antibody produced in mouse, clone 1H3, ascites fluid
SAB5300346
Monoclonal Anti-ALCAM antibody produced in mouse, clone 3F8B12, ascites fluid
SAB5300347 Monoclonal Anti-ALCAM antibody produced in mouse, clone 10F1G12, ascites fluid
SAB4700260
Monoclonal Anti-CD105-FITC antibody produced in mouse, clone MEM-226, purified immunoglobulin, buffered aqueous solution
SAB4700263 Monoclonal Anti-CD105-FITC antibody produced in mouse, clone MEM-229, purified immunoglobulin, buffered aqueous solution
AMAB90897
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1637, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90898
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1638, purified immunoglobulin, buffered aqueous glycerol solution
C7673
Monoclonal Anti-CD14 antibody produced in mouse, clone UCHM-1, purified immunoglobulin, buffered aqueous solution
SAB4700097
Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
SAB4700103 Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700669 Monoclonal Anti-CD14 antibody produced in mouse, clone B-A8, purified immunoglobulin, buffered aqueous solution
WH0000947M1
Monoclonal Anti-CD34 antibody produced in mouse, clone 5F3, purified immunoglobulin, buffered aqueous solution
SAB1403649
Monoclonal Anti-CD34 antibody produced in mouse, clone 5A6, purified immunoglobulin, buffered aqueous solution
SAB4700160 Monoclonal Anti-CD34 antibody produced in mouse, clone 4H11[APG], purified immunoglobulin, buffered aqueous solution
SAB4700736 Monoclonal Anti-CD34 antibody produced in mouse, clone 581, purified immunoglobulin, buffered aqueous solution