PloS one
Identification of Toll-like receptor 9 as parapoxvirus ovis-sensing receptor in plasmacytoid dendritic cells.
PMID 25171368
Abstract
Parapoxvirus ovis (PPVO) is known for its immunostimulatory capacities and has been successfully used to generate vector vaccines effective especially in non-permissive host species. Murine conventional and plasmacytoid dendritic cells (cDC and pDC) are able to recognize PPVO. The PPVO-sensing receptor on pDC is hitherto unknown. In this study we aimed to define the pattern recognition receptor responsible for the activation of murine pDC by inactivated and replication-competent PPVO. We show that PPVO-induced expression of type I and type III interferons, pro-inflammatory cytokines, and co-stimulatory CD86 by bone marrow-derived pDC but not cDC is blocked by chloroquine, an inhibitor of endosomal maturation. The activation of pDC is independent of viral replication and depends mainly on TLR9. Moreover, the use of phosphatidylinositol 3-kinase inhibitor wortmannin or C-Jun-N-terminal kinase inhibitor SP600125 results in significant reduction of PPVO-induced pDC activation. Taken together, our data identify endosomal TLR9 as PPVO-sensing receptor in pDC.