EMAIL THIS PAGE TO A FRIEND
Vaccine
Preparation and testing of a Vi conjugate vaccine using pneumococcal surface protein A (PspA) from Streptococcus pneumoniae as the carrier protein.
PMID 25171842
Abstract
In the current study pneumococcal surface protein A (PspA) was conjugated to Vi capsular polysaccharide from Salmonella Typhi to make available a vaccine against typhoid fever that has the potential to also provide broad protection from Streptococcus pneumoniae. High yielding production processes were developed for the purification of PspAs from families 1 and 2. The purified PspAs were conjugated to Vi with high recovery of both Vi and PspA. The processes developed especially for PspA family 2 could readily be adapted for large scale production under cGMP conditions. Previously we have shown that conjugation of diphtheria toxoid (DT) to Vi polysaccharide improves the immune response to Vi but can also enhance the response to DT. In this study it was shown that conjugation of PspA to Vi enhanced the anti-PspA response and that PspA was a suitable carrier protein as demonstrated by the characteristics of a T-cell dependent response to the Vi. We propose that a bivalent vaccine consisting of PspA from families 1 and 2 bound to Vi polysaccharide would protect against typhoid fever and has the potential to also protect against pneumococcal disease and should be considered for use in developing countries.