EMAIL THIS PAGE TO A FRIEND
FEBS letters
MicroRNA-29a suppresses the growth, migration, and invasion of lung adenocarcinoma cells by targeting carcinoembryonic antigen-related cell adhesion molecule 6.
PMID 25171863
Abstract
Carcinoembryonic antigen-related cell adhesion molecule 6 (CEACAM6) is an important regulator of cell adhesion, invasion, and metastasis. The aim of this study was to evaluate the functional roles of CEACAM6 in lung adenocarcinoma and to identify miRNAs that inhibit the growth, migration, and invasion of lung adenocarcinoma cells by targeting CEACAM6. CEACAM6 expression is associated with poor prognosis of patients with lung adenocarcinoma, and CEACAM6 has important functional roles in controlling the growth, migration, and invasion of lung adenocarcinoma cells in vitro and in vivo. Furthermore, miR-29a can suppress the growth, migration, and invasion of lung adenocarcinoma cells by targeting CEACAM6. Therefore, miR-29a/CEACAM6 axis represents a potential therapeutic target for treatment of lung adenocarcinoma.