EMAIL THIS PAGE TO A FRIEND
Food chemistry
Antioxidant and cytoprotective properties of partridgeberry polyphenols.
PMID 25172753
Abstract
Partridgeberry (Vaccinium vitis-idaea) is a polyphenol-rich berry of the Ericaceous family, grown in Newfoundland and Labrador province of Canada. The aims of this study were to identify extraction solvents for the maximum recovery of polyphenols, to establish fractionation technique for isolation of major sub-classes of polyphenols, and to evaluate antioxidant and cytoprotective properties of the partridgeberry polyphenol preparations. The acidified 70% acetone was identified as the ideal solvent for the maximum recovery of polyphenols from partridgeberry. Further, aqueous two-phase extraction, column chromatography and UPLC-MS/MS were employed to produce three partridgeberry polyphenol fractions, rich in either, anthocyanins, flavan-3-ols or flavonols. All the three PPF were potent antioxidants and displayed cytoprotective activity through the activation of nuclear factor erythroid 2-related factor 2 pathway, scavenging of reactive oxygen species, and inhibition of cellular death. The current study suggests that partridgeberry has numerous potential health implications in both prevention and amelioration of various diseases involving oxidative stress.