EMAIL THIS PAGE TO A FRIEND
Analytica Chimica Acta
Pressurized liquid extraction and liquid chromatography-tandem mass spectrometry applied to determine iodinated X-ray contrast media in sewage sludge.
PMID 25172819