EMAIL THIS PAGE TO A FRIEND
Cancer research
Stress signaling from human mammary epithelial cells contributes to phenotypes of mammographic density.
PMID 25172842
Abstract
Telomere malfunction and other types of DNA damage induce an activin A-dependent stress response in mortal nontumorigenic human mammary epithelial cells that subsequently induces desmoplastic-like phenotypes in neighboring fibroblasts. Some characteristics of this fibroblast/stromal response, such as reduced adipocytes and increased extracellular matrix content, are observed not only in tumor tissues but also in disease-free breast tissues at high risk for developing cancer, especially high mammographic density tissues. We found that these phenotypes are induced by repression of the fatty acid translocase CD36, which is seen in desmoplastic and disease-free high mammographic density tissues. In this study, we show that epithelial cells from high mammographic density tissues have more DNA damage signaling, shorter telomeres, increased activin A secretion and an altered DNA damage response compared with epithelial cells from low mammographic density tissues. Strikingly, both telomere malfunction and activin A expression in epithelial cells can repress CD36 expression in adjacent fibroblasts. These results provide new insights into how high mammographic density arises and why it is associated with breast cancer risk, with implications for the definition of novel invention targets (e.g., activin A and CD36) to prevent breast cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-636
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, Anti-phospho-Histone H2A.X (Ser139), clone JBW301 is a well published Mouse Monoclonal Antibody validated in ChIP, ICC, IF, WB. This purified mAb is highly specific for phospho-Histone H2A.X (Ser139) also known as H2AXS139p.
HPA002018
Anti-CD36 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV48129
Anti-CD36 antibody produced in rabbit, affinity isolated antibody
SAB4300514
Anti-H2AFX (Ab-139) antibody produced in rabbit, affinity isolated antibody
SAB3500048
Anti-SCARB1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700477
Anti-SCARB1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107764
Anti-SCARB1 antibody produced in rabbit, affinity isolated antibody
PLA0294
Goat anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB4700165 Monoclonal Anti-CD36 antibody produced in mouse, clone TR9, purified immunoglobulin, buffered aqueous solution
SAB4700166 Monoclonal Anti-CD36 antibody produced in mouse, clone CB38 (NL07), purified immunoglobulin, buffered aqueous solution
WH0003014M5
Monoclonal Anti-H2AFX antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
SAB1403902 Monoclonal Anti-H2AFX, (N-terminal) antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
PLA0023
Rabbit anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.