EMAIL THIS PAGE TO A FRIEND
Molecular Cancer Therapeutics
Targeted silencing of MLL5? inhibits tumor growth and promotes gamma-irradiation sensitization in HPV16/18-associated cervical cancers.
PMID 25172963