EMAIL THIS PAGE TO A FRIEND
Journal of the neurological sciences
Tumor necrosis factor beta NcoI polymorphism is associated with inflammatory and metabolic markers in multiple sclerosis patients.
PMID 25173940
Abstract
To evaluate the association between the tumor necrosis factor beta (TNF-β) NcoI polymorphism and inflammatory and metabolic markers in patients with multiple sclerosis (MS) patients and the association of these markers with disease disability, a 782 base-pair fragment of the TNF-β gene was amplified from genomic DNA and digested with the NcoI restriction enzyme. The serum levels of numerous cytokines (IL-1β, IL-12, IL-6, TNF-α, IFN-γ, IL-4, IL-10, and IL-17) serum lipid levels, plasma insulin levels, and the Homeostasis Model Assessment-Insulin Resistance (HOMA-IR) levels were evaluated in 123 female and 43 male patients with MS. Females carrying the TNFB2/B2 genotype presented with decreased IL-4 and IL-10 levels and increased TNF-α, glucose, insulin, and HOMA-IR levels; moreover, there were positive correlations between EDSS and glucose and between EDSS and HOMA-IR in these females. Males carrying the TNFB2/B2 genotype exhibited increased levels of TNF-α, IFN-γ, and IL-17 (p=0.0326) and decreased levels of IL-4, IL-10, insulin, and HOMA-IR; there was a positive correlation between EDSS and TNF-α levels. The TNFB2/B2 genotype of TNF-β NcoI polymorphism was associated with increased inflammatory and metabolic markers and this association was different according to sex of MS patients.