EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Medicine
Sodium ferulate lowers portal pressure in rats with secondary biliary cirrhosis through the RhoA/Rho-kinase signaling pathway: a preliminary study.
PMID 25174394