EMAIL THIS PAGE TO A FRIEND
European Journal of Immunology
An amphioxus gC1q protein binds human IgG and initiates the classical pathway: Implications for a C1q-mediated complement system in the basal chordate.
PMID 25174509