EMAIL THIS PAGE TO A FRIEND
Nanoscale
Emission tunable, cyto/hemocompatible, near-IR-emitting Ag2S quantum dots by aqueous decomposition of DMSA.
PMID 25175037
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W318000
2-Mercaptopropionic acid, ≥95%, FG
C3H6O2S
D7881
meso-2,3-Dimercaptosuccinic acid, ~98%
C4H6O4S2