EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Effects of silica and calcium levels in nanobioglass ceramic particles on osteoblast proliferation.
PMID 25175236
Abstract
At nanoscale, bioglass ceramic (nBGC) particles containing calcium oxide (lime), silica and phosphorus pentoxide promote osteoblast proliferation. However, the role of varied amounts of calcium and silica present in nBGC particles on osteoblast proliferation is not yet completely known. Hence, the current work was aimed at synthesizing two different nBGC particles with varied amounts of calcium oxide and silica, nBGC-1: SiO2:CaO:P2O5; mol%~70:25:5 and nBGC-2: SiO2:CaO:P2O5; mol%~64:31:5, and investigating their role on osteoblast proliferation. The synthesized nBGC particles were characterized by transmission electron microscopy (TEM), energy dispersive X-ray spectroscopy (EDS), Fourier transform infrared (FTIR) spectroscopy and X-ray diffraction (XRD) studies. They exhibited their size at nanoscale and were non-toxic to human osteoblastic cells (MG-63). The nBGC-2 particles were found to have more effect on stimulation of osteoblast proliferation and promoted entering of more cells into G2/M cell cycle phase compared to nBGC-1 particles. There was a differential expression of cyclin proteins in MG-63 cells by nBGC-1 and nBGC-2 treatments, and the expression of cyclin B1 and E proteins was found to be more by nBGC-2 treatment. Thus, these results provide us a new insight in understanding the design of various nBGC particles by altering their ionic constituents with desirable biological properties thereby supporting bone augmentation.