EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
Antitumor effect of a copper (II) complex of a coumarin derivative and phenanthroline on lung adenocarcinoma cells and the mechanism of action.
PMID 25176185