EMAIL THIS PAGE TO A FRIEND
Nature Communications
A general and scalable synthesis approach to porous graphene.
PMID 25178835
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277908
Ammonium molybdate, 99.98% trace metals basis
H8MoN2O4
A1343
Ammonium molybdate, meets USP testing specifications
H24Mo7N6O24 · 4H2O
431346
Ammonium molybdate tetrahydrate, ACS reagent, 99.98% trace metals basis
H24Mo7N6O24 · 4H2O
A7302
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
M0878
Ammonium molybdate tetrahydrate, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
M1019
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
09878
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
H24Mo7N6O24 · 4H2O
38835
Bromide standard concentrate 10.00 gr Br-, analytical standard, 10.00 g/L, for 1 l standard solution
BrK
234761
Molybdenum(IV) oxide, 99%
MoO2
711209
Molybdenum(IV) oxide, 97%
MoO2
203815
Molybdenum(VI) oxide, 99.97% trace metals basis
MoO3
267856
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
MoO3
M0753
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%
MoO3
69850
Molybdenum(VI) oxide, puriss. p.a., 99.5%
MoO3
775703
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
MoO3
746444
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
BrK
793604
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
BrK
P2097
Potassium permanganate, meets USP testing specifications
KMnO4
207985
Potassium permanganate, 97%
KMnO4
60459
Potassium permanganate, BioUltra, ≥99.0% (RT)
KMnO4
60467
Potassium permanganate, tested according to Ph.Eur.
KMnO4
243655
Sodium molybdate, ≥98%
MoNa2O4
737860
Sodium molybdate, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
MoNa2O4
14505
Sodium stannate, 42-45% SnO2 basis
Na2O3Sn