EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Evaluation of quadrupole-time-of-flight mass spectrometry in comprehensive two-dimensional gas chromatography.
PMID 25179286
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M47949
2-Methylheptane, 98%
C8H18
185841
Dicyclohexylamine, 99%
C12H23N
442564
Dicyclohexylamine, analytical standard
C12H23N
Y0000279
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard
C12H27O4P
00675
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)
C12H27O4P
90820
Tributyl phosphate, puriss., ≥99.0% (GC)
C12H27O4P
158615
Tributyl phosphate, 97%
C12H27O4P
240494
Tributyl phosphate, ≥99%
C12H27O4P
34108
Tributyl phosphate, analytical standard
C12H27O4P
PHR1205
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
C12H27O4P
241288
Triphenyl phosphate, ≥99%
C18H15O4P
442829
Triphenyl phosphate, analytical standard
C18H15O4P
551392
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.
C18H15O4P
611735
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
C18H15O4P
48064
Triphenyl phosphate solution, certified reference material, 500 μg/mL in methyl tert-butyl ether
C18H15O4P
708968
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D)
C18H15O4P
733792
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
C18H15O4P
733741
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
C18H15O4P
736368
Triphenyl phosphate solution, NMR reference standard, 0.0485 M in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
C18H15O4P
756911
Triphenyl phosphate solution, NMR reference standard, 3 mM in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.
C18H15O4P