EMAIL THIS PAGE TO A FRIEND
Transplant International : Official Journal of the European Society For Organ Transplantation
Renal dysfunction and intragraft proMMP9 activity in renal transplant recipients with interstitial fibrosis and tubular atrophy.
PMID 25179305