EMAIL THIS PAGE TO A FRIEND
Environmental Toxicology and Pharmacology
Association of inflammatory response and oxidative injury in the pathogenesis of liver steatosis and insulin resistance following subchronic exposure to malathion in rats.
PMID 25180440