EMAIL THIS PAGE TO A FRIEND
Cell Death & Disease
Targeting Btk/Etk of prostate cancer cells by a novel dual inhibitor.
PMID 25188519
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
270458
1-Methyl-2-pyrrolidinone, for HPLC, ≥99%
C5H9NO
43729
1-Methyl-2-pyrrolidinone, for metal speciation analysis, ≥99.0% (GC)
C5H9NO
M6762
1-Methyl-2-pyrrolidinone
C5H9NO
328634
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
C5H9NO
494496
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
C5H9NO
242799
1-Methyl-2-pyrrolidinone, spectrophotometric grade, ≥99%
C5H9NO
443778
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
C5H9NO
78769
1-Methyl-2-pyrrolidinone, analytical standard
C5H9NO
69118
1-Methyl-2-pyrrolidinone, puriss. p.a., ≥99.0% (GC)
C5H9NO
M79603
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
C5H9NO
CDS000967
4-(4-Pyridyl)benzoic acid, AldrichCPR
C12H9NO2
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
00712
Acetonitrile solution, ~20% in H2O, for protein sequence analysis
C2H3N
516961
Ammonium formate, ≥99.995% trace metals basis
CH5NO2
09735
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
CH5NO2
70221
Ammonium formate, for mass spectrometry, ≥99.0%
CH5NO2
78314
Ammonium formate solution, BioUltra, 10 M in H2O
CH5NO2
714690
Ammonium formate solution, 10 mM in H2O, for HPLC
CH5NO2
481432
Cyanogen bromide, 99.995% trace metals basis
CBrN
16772
Cyanogen bromide, ≥98.5% (RT)
CBrN
C91492
Cyanogen bromide, reagent grade, 97%
CBrN
261610
Cyanogen bromide solution, 5.0 M in acetonitrile
CBrN
341894
Cyanogen bromide solution, 3.0 M in methylene chloride
CBrN
L1002
L-Leucine methyl ester hydrochloride, 98%
C7H15NO2 · HCl
387649
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
C8H19N
496219
N,N-Diisopropylethylamine, 99.5%, biotech. grade
C8H19N
D125806
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
C8H19N
694134
N-(3-Aminopropyl)piperidine, 95%
C8H18N2
03440
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
C8H19N