EMAIL THIS PAGE TO A FRIEND
Canadian Journal of Physiology and Pharmacology
Distribution of aquaporin genes and selection of individual reference genes for quantitative real-time RT-PCR analysis in multiple tissues of the mouse.
PMID 25188728