EMAIL THIS PAGE TO A FRIEND
International journal of oncology
MicroRNA-135b promotes proliferation, invasion and migration of osteosarcoma cells by degrading myocardin.
PMID 25190111
Abstract
MicroRNA-135b (miR‑135b) functions as an oncogene in malignant tumors suggesting that it might be a target for malignant tumor therapy, but its role in osteosarcoma has not been investigated to date. In this study, we found that myocardin expression is specifically attenuated in MG63 osteosarcoma cells. Silencing miR‑135b restores the expression of myocardin and miR‑135b inhibits the expression of myocardin by targeting its 3'UTR in MG63 osteosarcoma cells. In additional experiments, we found that miR‑135b was highly expressed in osteosarcoma tissues and it promoted MG63 cell proliferation, migration and invasion. Myocardin had the opposite effects of miR‑135b, which suppressed proliferation, migration and invasion in MG63 cells. Thus, it is probably that miR‑135b promotes proliferation, invasion and migration of osteosarcoma cells by degrading myocardin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2502098
Anti-PCNA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA030522
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA030523
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
HPA030521
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
SAB4502103
Anti-PCNA antibody produced in rabbit, affinity isolated antibody
SAB2701819
Anti-PCNA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
WH0005111M2
Monoclonal Anti-PCNA antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution
SAB1404178
Monoclonal Anti-PCNA antibody produced in mouse, clone S1, purified immunoglobulin, buffered aqueous solution
SAB4100556
Monoclonal Anti-PCNA antibody produced in mouse, clone PCNA -A509, culture supernatant
PLA0079
Rabbit anti-PCNA Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0080
Rabbit anti-PCNA Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB2502099
Anti-PCNA antibody produced in goat, affinity isolated antibody, buffered aqueous solution