EMAIL THIS PAGE TO A FRIEND
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology
Time-kill effect of levofloxacin on multidrug-resistant Pseudomonas aeruginosa and Acinetobacter baumannii: synergism with imipenem and colistin.
PMID 25192733
Abstract
In the present study, we challenged the concept that levofloxacin should not be used for the management of ventilator-associated pneumonia (VAP) when minimum inhibitory concentrations (MICs) exceed 2 μg/ml. Multidrug-resistant (MDR) and genetically distinct isolates of Pseudomonas aeruginosa (n = 49) and Acinetobacter baumannii (n = 29) from patients with VAP were exposed over time to levofloxacin, imipenem, colistin and their combinations. Synergy between levofloxacin and imipenem was found in 55.3 % and between levofloxacin and colistin in 90.9 % of isolates of P. aeruginosa within the first 4 h of growth. Synergy with imipenem but not with colistin was dependent of the MIC. Synergy between levofloxacin and imipenem was found in 58.6 % of isolates of A. baumannii after 24 h of growth. Considerable synergy was found between levofloxacin and colistin, reaching 84.8 % of isolates of A.baumannii after 6 h of growth. Synergy was independent from the MIC. These results create hopes that levofloxacin can be used as combination therapy for infections by MDR bacteria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1362103
Levofloxacin, United States Pharmacopeia (USP) Reference Standard
C18H20FN3O4 · 0.5H2O
40922
Levofloxacin, analytical standard
C18H20FN3O4