EMAIL THIS PAGE TO A FRIEND
The AAPS journal
Structure, size, and solubility of antigen arrays determines efficacy in experimental autoimmune encephalomyelitis.
PMID 25193268
Abstract
Presentation of antigen with immune stimulating "signal" has been a cornerstone of vaccine design for decades. Here, the antigen plus immune "signal" of vaccines is modified to produce antigen-specific immunotherapies (antigen-SITs) that can potentially reprogram the immune response toward tolerance of an autoantigen. The codelivery of antigen with a cell adhesion inhibitor using Soluble Antigen Arrays (SAgAs) was previously shown to slow or halt experimental autoimmune encephalomyelitis (EAE), a murine form of multiple sclerosis (MS). SAgAs are comprised of a hyaluronic acid backbone with cografted intercellular cell adhesion molecule-1 ligand derived from αL-integrin (CD11a237-246, "LABL") and an encephalitogenic epitope peptide of proteolipid protein (PLP139-151, "PLP"). Here, the physical characteristics of the carrier were investigated to evaluate how structure, size, and solubility drive the immune response when treating EAE. A bifunctional peptide (small, soluble), SAgAs (large, soluble), and PLGA nanoparticles (large, insoluble) all displaying PLP and LABL in equimolar ratios were compared. Maximum EAE suppression was achieved with coincident display of both peptides on a soluble construct.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71251
Acetic acid, analytical standard
C2H4O2
90872
Acetone, analytical standard
C3H6O
1006801
Acetone, United States Pharmacopeia (USP) Reference Standard
C3H6O
PHR1219
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
C3H6O
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
90577
Bifido Selective Supplement B, for microbiology
C2H4O2
M9546
D-Mannitol, BioXtra, ≥98% (HPLC)
C6H14O6
M9647
D-Mannitol, ACS reagent
C6H14O6
M4125
D-Mannitol, ≥98%
C6H14O6
M1902
D-Mannitol, plant cell culture tested
C6H14O6
M8429
D-Mannitol, meets EP, FCC, USP testing specifications
C6H14O6
63559
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
C6H14O6
63560
D-Mannitol, ACS reagent, for microbiology, ≥99.0%
C6H14O6
17311
D-Mannitol, tested according to Ph.Eur.
C6H14O6
78513
D-Mannitol, ≥99.9999% (metals basis), for boron determination
C6H14O6
270997
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
CH2Cl2
270563
Dichloromethane, ≥99.9%
CH2Cl2
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
48538
Dichloromethane, analytical standard, stabilized
CH2Cl2
07-2205
Dichloromethane, suitable for PCB analysis
CH2Cl2
07-2206
Dichloromethane, JIS 5000, for residue analysis
CH2Cl2
07-2230
Dichloromethane, for HPLC
CH2Cl2
48600
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
CH2Cl2
40042
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
CH2Cl2
723851
Dichloromethane solution, contains 10 % (v/v) methanol
CH2Cl2
296082
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
C4H10O
91238
Diethyl ether, analytical standard
C4H10O
31700
Diethyl ether, purum, ≥99.0% (GC)
C4H10O