EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Use of mep HyperCel for polishing of human serum albumin.
PMID 25195026
Abstract
The manufacture of human serum albumin by chromatographic procedures involves gel filtration chromatography as a final polishing step. Despite this step being essential to remove high molecular weight impurity proteins and thus ensure a stable and safe final product, it is relatively inefficient. This paper explores the use of hydrophobic charge induction chromatographic media, MEP HyperCel as an alternative to Sephacryl S200HR gel filtration for the polishing of human serum albumin derived by ion exchange chromatographic purification of Cohn Supernatant I. The use of MEP HyperCel results in a product with a higher purity than achieved with gel filtration and in a less time consuming manner and with potential resource savings. MEP HyperCel appears to have great potential for incorporation into downstream processes in the plasma fractionation industry as an efficient means of achieving polishing of intermediates or capture of proteins of interest.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471321
2-(Diethylamino)ethanol, ≥99.5%
C6H15NO