EMAIL THIS PAGE TO A FRIEND
Targeted diagnosis and therapy
Immobilization of antigens and antibodies on chromium dioxide magnetic particles for use in immunodiagnostic assays.
PMID 2519547
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480037
Magtrieve
CrO2