EMAIL THIS PAGE TO A FRIEND
Journal of labelled compounds & radiopharmaceuticals
Synthesis of carbon-14 and stable isotope labeled Avagacestat: a novel gamma secretase inhibitor for the treatment of Alzheimer's disease.
PMID 25196195
Abstract
Bristol-Myers Squibb and others are developing drugs that target novel mechanisms to combat Alzheimer's disease. γ-Secretase inhibitors are one class of potential therapies that have received considerable attention. (R)-2-(4-Chloro-N-(2-fluoro-4-(1,2,4-oxadiazol-3-yl)benzyl)phenylsulfonamido)-5,5,5-trifluoropentanamide (Avagacestat) is a γ-secretase-inhibiting drug that has been investigated by Bristol-Myers Squibb in preclinical and clinical studies. An important step in the development process was the synthesis of a carbon-14-labeled analog for use in a human absorption, distribution, metabolism, and excretion study and a stable isotope labeled analog for use as a standard in bioanalytical assays to accurately quantify the concentration of the drug in biological samples. Carbon-14 labeled Avagacestat was synthesized in seven steps in a 33% overall yield from carbon-14 labeled potassium cyanide. A total of 5.95 mCi was prepared with a specific activity of 0.81 μCi/mg and a radiochemical purity of 99.9%. (13) C6 -Labeled Avagacestat was synthesized in three steps in a 15% overall yield from 4-chloro[(13) C6 ]aniline. A total of 585 mg was prepared with a ultraviolet purity of 99.9%.