EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
Design and evaluation of a novel trifluorinated imaging agent for assessment of bile acid transport using fluorine magnetic resonance imaging.
PMID 25196788