EMAIL THIS PAGE TO A FRIEND
Cancer letters
SMILE upregulated by metformin inhibits the function of androgen receptor in prostate cancer cells.
PMID 25199764
Abstract
Metformin, a diabetes drug, has been reported to inhibit the growth of prostate cancer cells. In this study, we investigated the effect and action mechanism of metformin on the function of androgen receptor (AR), a key molecule in the proliferation of prostate cancer cells. Metformin was found to reduce androgen-dependent cell growth and the expression of AR target genes by inhibiting AR function in prostate cancer cells such as LNCaP and C4-2 cells. Interestingly, metformin upregulated the protein level of small heterodimer partner-interacting leucine zipper (SMILE), a coregulator of nuclear receptors, and knockdown of SMILE expression with shRNA abolished the inhibitory effect of metformin on AR function. Further studies revealed that SMILE protein itself suppressed the transactivation of AR, and its ectopic expression resulted in the repressed expression of endogenous AR target genes, PSA and NKX3.1, in LNCaP cells. In addition, SMILE protein physically interacted with AR and competed with the AR coactivator SRC-1 to modulate AR transactivation. As expected, SMILE repressed androgen-dependent growth of LNCaP and C4-2 cells. Taken together, these results suggest that SMILE, which is induced by metformin, functions as a novel AR corepressor and may mediate the inhibitory effect of metformin on androgen-dependent growth of prostate cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1300902
Anti-APS (center) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1405474
Anti-KLK3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000764
Anti-KLK3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410703
Anti-KLK3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501649
Anti-KLKB1 (heavy chain, Cleaved-Arg390) antibody produced in rabbit, affinity isolated antibody
HPA005634
Anti-KLKB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV45678
Anti-KLKB1 antibody produced in rabbit, IgG fraction of antiserum
SAB2106715
Anti-KLKB1 antibody produced in rabbit, affinity isolated antibody
SAB4501766
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501767
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501764
Anti-Lamin A antibody produced in rabbit, affinity isolated antibody
SAB1406087
Anti-LMNA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006660
Anti-LMNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501531
Anti-Prostate-specific Antigen antibody produced in rabbit, affinity isolated antibody
WH0000354M2
Monoclonal Anti-KLK3 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
SAB5300268
Monoclonal Anti-KLK3 antibody produced in mouse, clone 5A11E9, ascites fluid
SAB4200236
Monoclonal Anti-Lamin A/C antibody produced in mouse, clone 4C11, purified from hybridoma cell culture