EMAIL THIS PAGE TO A FRIEND
Investigational New Drugs
Momordica charantia lectin exhibits antitumor activity towards hepatocellular carcinoma.
PMID 25200916
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0186
BI-6C9, ≥97% (HPLC), solid
C23H25N3O4S2
S8307
SB 203580, solid, ≥98% (HPLC)
C21H16FN3OS