EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Aptamer-functionalized solid phase microextraction-liquid chromatography/tandem mass spectrometry for selective enrichment and determination of thrombin.
PMID 25201271
Abstract
In this publication, a novel solid phase microextraction (SPME) coating functionalized with a DNA aptamer for selective enrichment of a low abundance protein from diluted human plasma is described. This approach is based on the covalent immobilization of an aptamer ligand on electrospun microfibers made with the hydrophilic polymer poly(acrylonitrile-co-maleic acid) (PANCMA) on stainless steel rods. A plasma protein, human α-thrombin, was employed as a model protein for selective extraction by the developed Apt-SPME probe, and the detection was carried out with liquid chromatography/tandem mass spectrometry (LC-MS/MS). The SPME probe exhibited highly selective capture, good binding capacity, high stability and good repeatability for the extraction of thrombin. The protein selective probe was employed for direct extraction of thrombin from 20-fold diluted human plasma samples without any other purification. The Apt-SPME method coupled with LC-MS/MS provided a good linear dynamic range of 0.5-50 nM in diluted human plasma with a good correlation coefficient (R(2)=0.9923), and the detection limit of the proposed method was found to be 0.30 nM. Finally, the Apt-SPME coupled with LC-MS/MS method was successfully utilized for the determination of thrombin in clinical human plasma samples. One shortcoming of the method is its reduced efficiency in undiluted human plasma compared to the standard solution. Nevertheless, this new aptamer affinity-based SPME probe opens up the possibility of selective enrichment of a given targeted protein from complex sample either in vivo or ex vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31564
Acrylonitrile, analytical standard
C3H3N
110213
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
C3H3N
320137
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
C3H3N
PHR1553
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO
63200
Maleic anhydride, puriss., ≥99.0% (NT)
C4H2O3
M188
Maleic anhydride, 99%
C4H2O3
M625
Maleic anhydride, powder, 95%
C4H2O3
72438
N,N-Dimethylformamide, analytical standard
C3H7NO
56480
N-Hydroxysuccinimide, purum, ≥97.0% (T)
C4H5NO3
60489
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
K2O8S2
379824
Potassium persulfate, 99.99% trace metals basis
K2O8S2
216224
Potassium persulfate, ACS reagent, ≥99.0%
K2O8S2
S0505
Sodium sulfite, ≥98%
Na2O3S
71988
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)
Na2O3S
31454
Sodium sulfite, puriss. p.a., Reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)
Na2O3S
13471
Sodium sulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, anhydrous, 95.0-100.0% (iodometric)
Na2O3S
08981
Sodium sulfite, analytical standard
Na2O3S
239321
Sodium sulfite, ACS reagent, ≥98.0%
Na2O3S
S4672
Sodium sulfite, BioXtra, ≥98%
Na2O3S
901916
Sodium sulfite, anhydrous, Redi-Dri, reagent grade, ≥98%
Na2O3S