EMAIL THIS PAGE TO A FRIEND
The Journal of Steroid Biochemistry and Molecular Biology
Next-generation steroidogenesis inhibitors, dutasteride and abiraterone, attenuate but still do not eliminate androgen biosynthesis in 22RV1 cells in vitro.
PMID 25201454