EMAIL THIS PAGE TO A FRIEND
Anticancer Research
The multi-tyrosine kinase inhibitor TKI258, alone or in combination with RAD001, is effective for treatment of human leukemia with BCR-ABL translocation in vitro.
PMID 25202073