EMAIL THIS PAGE TO A FRIEND
Journal of Neurotrauma
Immunosuppression of allogenic mesenchymal stem cells transplantation after spinal cord injury improves graft survival and beneficial outcomes.
PMID 25203134