EMAIL THIS PAGE TO A FRIEND
Basic and Clinical Pharmacology and Toxicology
Cardioprotective and cardiotoxic effects of quercetin and two of its in vivo metabolites on differentiated h9c2 cardiomyocytes.
PMID 25203460