EMAIL THIS PAGE TO A FRIEND
Environmental Research
Bioavailability, oxidative stress, neurotoxicity and genotoxicity of pharmaceuticals bound to marine sediments. The use of the polychaete Hediste diversicolor as bioindicator species.
PMID 25203819