EMAIL THIS PAGE TO A FRIEND
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association
Omega-3 fatty acids inhibit the up-regulation of endothelial chemokines in maintenance hemodialysis patients.
PMID 25204316
Abstract
Chronic systemic inflammation is common in patients with chronic kidney disease on dialysis (CKD5D) and has been considered a key mediator of the increased cardiovascular risk in this patient population. In this study, we tested the hypothesis that supplementation of omega-3 polyunsaturated fatty acids (ω-3 PUFAs) will attenuate the systemic inflammatory process in CKD5D patients. The design was a randomized, double-blinded, placebo controlled pilot trial (NCT00655525). Thirty-eight patients were randomly assigned in a 1 : 1 fashion to receive 2.9 g of eicosapentaenoic acid (C20:5, n-3) plus docosahexaenoic acid (C22:6, n-3) versus placebo for 12 weeks. The primary outcome was change in pro-inflammatory chemokines measured by lipopolysaccharide (LPS)-stimulated peripheral blood mononuclear cells (PBMCs). Secondary outcomes were changes in systemic inflammatory markers. Analysis of covariance was used to compare percent change from baseline to 12 weeks. Thirty-one patients completed 12 weeks and three patients completed 6 weeks of the study. Median age was 52 (interquartile range 45, 60) years, 74% were African-American and 79% were male. Supplementation of ω-3 PUFAs effectively decreased the LPS-induced PBMC expression of RANTES (Regulated upon Activation, Normal T cell Expressed and Secreted) and MCP-1 (Monocyte Chemotactic Protein-1; unadjusted P = 0.04 and 0.06; adjusted for demographics P = 0.02 and 0.05, respectively). There was no significant effect of the intervention on serum inflammatory markers (C-reactive protein, interleukin-6 and procalcitonin). The results of this pilot study suggest that supplementation of ω-3 PUFAs is beneficial in decreasing the levels of endothelial chemokines, RANTES and MCP-1. Studies of larger sample size and longer duration are required to further evaluate effects of ω-3 PUFAs on systemic markers of inflammation, other metabolic parameters and clinical outcomes, particularly cardiovascular outcomes in CKD5D patients.