EMAIL THIS PAGE TO A FRIEND
Acta Physiologica (Oxford, England)
Late gestation under- and overnutrition have differential impacts when combined with a post-natal obesogenic diet on glucose-lactate-insulin adaptations during metabolic challenges in adolescent sheep.
PMID 25204637