EMAIL THIS PAGE TO A FRIEND
Multiple sclerosis (Houndmills, Basingstoke, England)
ACTRIMS-ECTRIMS MSBoston 2014: Poster Sessions 2.
PMID 25205049
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU072881 MISSION® esiRNA, esiRNA targeting mouse 1190002H23Rik (esiRNA1)