EMAIL THIS PAGE TO A FRIEND
Circulation
?-adrenergic receptor-mediated cardiac contractility is inhibited via vasopressin type 1A-receptor-dependent signaling.
PMID 25205804
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V9879
[Arg8]-Vasopressin acetate salt, ≥95% (HPLC)
C46H65N15O12S2 · xC2H4O2