EMAIL THIS PAGE TO A FRIEND
The Journal of Investigative Dermatology
Peripheral and local human papillomavirus 16-specific CD8+ T-cell expansions characterize erosive oral lichen planus.
PMID 25207820