EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Factors affecting the adsorptive removal of bisphenol A in landfill leachate by high silica Y-type zeolite.
PMID 25209542
Abstract
Although bisphenol A (BPA), a representative endocrine-disrupting compound, has been detected frequently in landfill leachate, effective technologies for BPA removal from landfill leachates are limited. We used high silica Y-type zeolite (HSZ-385) for the selective adsorption of BPA from landfill leachate, and factors affecting this adsorption are discussed. Higher removal efficiencies at pH 5.0-9.0 imply that neutral BPA is adsorbed more easily onto HSZ-385 than monomeric or divalent BPA anions. An increase in ionic strength and sodium acetate concentration did not affect BPA adsorption significantly, while the removal efficiency decreased slightly when more than 50 mgC/L of humic acid was added. HSZ-385 was applied to synthetic leachates that simulate the composition of landfill leachate at various degradation stages. In young acidic leachates that contain sodium acetate, the use of HSZ-385 for the adsorptive removal of BPA appears to be more effective than in old alkaline leachates, which contain large amounts of humic acid. In addition, 82 % BPA removal was achieved from young raw leachates using HSZ-385, which demonstrates that selective BPA removal from actual landfill leachate has been achieved.