EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Renal Physiology
Sex-specific effect of antenatal betamethasone exposure on renal oxidative stress induced by angiotensins in adult sheep.
PMID 25209867