EMAIL THIS PAGE TO A FRIEND
BMC Infectious Diseases
Proliferative capacity and cytokine production by cells of HIV-infected and uninfected adults with different helminth infection phenotypes in South Africa.
PMID 25209883