EMAIL THIS PAGE TO A FRIEND
PLoS neglected tropical diseases
IL-10 and NOS2 modulate antigen-specific reactivity and nerve infiltration by T cells in experimental leprosy.
PMID 25210773
Abstract
Although immunopathology dictates clinical outcome in leprosy, the dynamics of early and chronic infection are poorly defined. In the tuberculoid region of the spectrum, Mycobacterium leprae growth is restricted yet a severe granulomatous lesion can occur. The evolution and maintenance of chronic inflammatory processes like those observed in the leprosy granuloma involve an ongoing network of communications via cytokines. IL-10 has immunosuppressive properties and IL-10 genetic variants have been associated with leprosy development and reactions. The role of IL-10 in resistance and inflammation in leprosy was investigated using Mycobacterium leprae infection of mice deficient in IL-10 (IL-10-/-), as well as mice deficient in both inducible nitric oxide synthase (NOS2-/-) and IL-10 (10NOS2-/-). Although a lack of IL-10 did not affect M. leprae multiplication in the footpads (FP), inflammation increased from C57Bl/6 (B6)