EMAIL THIS PAGE TO A FRIEND
American journal of otolaryngology
FoxP3 and indoleamine 2,3-dioxygenase immunoreactivity in sentinel nodes from melanoma patients.
PMID 25212103
Abstract
1) Assess FoxP3/indoleamine 2,3-dioxygenase immunoreactivity in head and neck melanoma sentinel lymph nodes and 2) correlate FoxP3/indoleamine 2,3-dioxygenase with sentinel lymph node metastasis and clinical recurrence. Retrospective cohort study. Patients with sentinel lymph node biopsy for head and neck melanoma between 2004 and 2011 were identified. FoxP3/indoleamine 2,3-dioxygenase prevalence and intensity were determined from the nodes. Poor outcome was defined as local, regional or distant recurrence. The overall immunoreactivity score was correlated with clinical recurrence and sentinel lymph node metastasis using the chi-square test for trend. Fifty-six sentinel lymph nodes were reviewed, with 47 negative and 9 positive for melanoma. Patients with poor outcomes had a statistically significant trend for higher immunoreactivity scores (p=0.03). Positive nodes compared to negative nodes also had a statistically significant trend for higher immunoreactivity scores (p=0.03). Among the negative nodes, there was a statistically significant trend for a poor outcome with higher immunoreactivity scores (p=0.02). FoxP3/indoleamine 2,3-dioxygenase immunoreactivity correlates with sentinel lymph node positivity and poor outcome. Even in negative nodes, higher immunoreactivity correlated with poor outcome. Therefore higher immunoreactivity may portend a worse prognosis even without metastasis in the sentinel lymph node. This could identify a subset of patients that may benefit from future trials and treatment for melanoma through Treg and IDO suppression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41764
Anti-GNAS (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV42694
Anti-GNAS (AB3) antibody produced in rabbit, IgG fraction of antiserum
HPA018122
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA028386
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA027478
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
SAB2100941
Anti-GNAS antibody produced in rabbit, affinity isolated antibody
SAB2501926
Anti-IDO1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA023072
Anti-IDO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA023149
Anti-IDO1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA027772
Anti-IDO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
SAB1406038
Anti-INDO antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411912 ANTI-INDO antibody produced in mouse, clone 4C9, purified immunoglobulin, buffered aqueous solution
SAB1411913
ANTI-INDO antibody produced in mouse, clone 4A6, purified immunoglobulin, buffered aqueous solution
SAB1411914
ANTI-INDO antibody produced in mouse, clone 4B1, purified immunoglobulin, buffered aqueous solution
SAB1411915
ANTI-INDO antibody produced in mouse, clone 4F9, purified immunoglobulin, buffered aqueous solution
SAB1411916
ANTI-INDO antibody produced in mouse, clone 1D5, purified immunoglobulin, buffered aqueous solution
SAB1411917
ANTI-INDO antibody produced in mouse, clone 1C5, purified immunoglobulin, buffered aqueous solution
SAB1411918
ANTI-INDO antibody produced in mouse, clone 1C1, purified immunoglobulin, buffered aqueous solution
HPA049079
Anti-LIPH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA021656
Anti-SLC10A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3701446
Monoclonal Anti-IDO1 antibody produced in mouse, clone 10.1, purified immunoglobulin
SAB1403979
Monoclonal Anti-INDO antibody produced in mouse, clone 3D12-C6, ascites fluid
WH0003620M5
Monoclonal Anti-INDO antibody produced in mouse, clone 4D2, purified immunoglobulin, buffered aqueous solution
AV41765
Anti-GNAS (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1400841
Anti-LIPH antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2105728
Anti-SLC10A3 antibody produced in rabbit, affinity isolated antibody
SAB2107647
Anti-LIPH antibody produced in rabbit, affinity isolated antibody