EMAIL THIS PAGE TO A FRIEND
The Journal of reproduction and development
Inhibitory effect of insulin-like growth factor-binding protein-7 (IGFBP7) on in vitro angiogenesis of vascular endothelial cells in the rat corpus luteum.
PMID 25212428
Abstract
Angiogenesis in the developing corpus luteum (CL) is a prerequisite for establishment and maintenance of an early pregnancy. To explore the physiological significance of insulin-like growth factor-binding protein-7 (IGFBP7) in the developing CL, the effects of IGFBP7 on vascular endothelial growth factor (VEGFA)- and luteinizing hormone (LH)-induced in vitro tube formation were tested using isolated luteal microvascular endothelial cells (LECs). Capillary-like tube formation of LECs and their proliferation were stimulated by both VEGFA and LH. IGFBP7 treatment suppressed VEGFA- or LH-induced tube formation. The proliferation and migration of LECs, and phosphorylation of mitogen-activated protein kinase kinase and extracellular signal-regulated kinase 1/2 were inhibited by IGFBP7. Furthermore, IGFBP7 attenuated VEGFA-enhanced cyclooxygenase (COX)-2 mRNA expression and prostaglandin E2 secretion. These findings suggest the possibility that luteal IGFBP7 secretion may suppress the stimulatory effect of VEGFA on angiogenesis in the early CL.