EMAIL THIS PAGE TO A FRIEND
The Journal of Infectious Diseases
Rifaximin has a marginal impact on microbial translocation, T-cell activation and inflammation in HIV-positive immune non-responders to antiretroviral therapy - ACTG A5286.
PMID 25214516