EMAIL THIS PAGE TO A FRIEND
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery
Intratympanic steroids as a salvage treatment for sudden sensorineural hearing loss? A meta-analysis.
PMID 25217083
Abstract
Sudden sensorineural hearing loss is typically treated with systemic steroids. The aim of this meta-analysis was to evaluate the efficacy of salvage intratympanic steroid treatment in patients who have initial treatment failure with systemic steroids. A MEDLINE literature search was performed, supported by searches of Web of Science, Biosis, and Science Direct. Articles of all languages were included. Selection of relevant publications was conducted independently by three authors. Only randomized controlled trials were considered. In one arm of the studies, the patients received salvage intratympanic steroids. In the other arm, patients did not receive further treatment. The standard difference in mean (SDM) amount of improvement in hearing threshold between patients who did and did not receive salvage intratympanic steroids was calculated. From an initial 184 studies found via the search strategy, 5 studies met inclusion criteria and were included. There was a statistically significant greater reduction in hearing threshold on pure-tone audiometry in patients who received salvage intratympanic steroids than in those who did not (SDM = -0.401, p = 0.005). Subgroup analysis showed that administration by intratympanic injection (SDM = -0.375, p = 0.013) rather than a round window catheter (SDM = -0.629, p = 0.160) yielded significant improvement in outcome. The usage of dexamethasone yielded better outcomes (SDM = -0.379, p = 0.039) than the use of methylprednisolone (SDM = -0.459, p = 0.187). No serious side effect of treatment was reported. In patients who have failed initial treatment with systemic steroids, additional treatment with salvage intratympanic dexamethasone injections demonstrate a statistically significant reduction in the hearing thresholds as compared to controls.