EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
An orphan esterase ABHD10 modulates probenecid acyl glucuronidation in human liver.
PMID 25217485
Abstract
Probenecid, a widely used uricosuric agent, is mainly metabolized to probenecid acyl glucuronide (PRAG), which is considered a causal substance of severe allergic or anaphylactoid reactions. PRAG can be hydrolyzed (deglucuronidated) to probenecid. The purpose of this study was to identify enzymes responsible for probenecid acyl glucuronidation and PRAG deglucuronidation in human livers and to examine the effect of deglucuronidation in PRAG formation. In human liver homogenates (HLHs), the intrinsic clearance (CLint) of PRAG deglucuronidation was much greater (497-fold) than that of probenecid acyl glucuronidation. Evaluation of PRAG formation by recombinant UDP-glucuronosyltransferase (UGT) isoforms and an inhibition study using HLHs as an enzyme source demonstrated that multiple UGT isoforms, including UGT1A1, UGT1A9, and UGT2B7, catalyzed probenecid acyl glucuronidation. We found that recombinant α/β hydrolase domain containing 10 (ABHD10) substantially catalyzed PRAG deglucuronidation activity, whereas carboxylesterases did not. Similar inhibitory patterns by chemicals between HLHs and recombinant ABHD10 supported the major contribution of ABHD10 to PRAG deglucuronidation in human liver. Interestingly, it was demonstrated that the CLint value of probenecid acyl glucuronidation in HLHs was increased by 1.7-fold in the presence of phenylmethylsulfonyl fluoride, which potently inhibited ABHD10 activity. In conclusion, we found that PRAG deglucuronidation catalyzed by ABHD10 suppressively regulates PRAG formation via multiple UGT enzymes in human liver. The balance of activities by these enzymes is important for the formation of PRAG, which may be associated with the adverse reactions observed after probenecid administration.