EMAIL THIS PAGE TO A FRIEND
Cancer Research
Targeted noninvasive imaging of EGFR-expressing orthotopic pancreatic cancer using multispectral optoacoustic tomography.
PMID 25217521
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2633
Cardiogreen, polymethine dye
C43H47N2NaO6S2
21980
Cardiogreen, for microscopy
C43H47N2NaO6S2